• USA Home
  • 529672 - Manganese(II) carbonate

EMAIL THIS PAGE TO A FRIEND
529672 Sigma-Aldrich

Manganese(II) carbonate

99.99% trace metals basis

Synonym: Manganese carbonate (MnCO3 ), Manganese carbonate

Purchase

Properties

assay   99.99% trace metals basis
form   powder
density   3.12 g/mL at 25 °C (lit.)
SMILES string   [Mn++].[O-]C([O-])=O
InChI   1S/CH2O3.Mn/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2
InChI key   XMWCXZJXESXBBY-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

377449 Manganese(II) carbonate, ≥99.9% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?