• USA Home
  • 529818 - Potassium hexachlorotellurate

EMAIL THIS PAGE TO A FRIEND
529818 Sigma-Aldrich

Potassium hexachlorotellurate

99.99% trace metals basis

Purchase

Properties

assay   99.99% trace metals basis
form   crystalline
SMILES string   [K+].[K+].Cl[Te--](Cl)(Cl)(Cl)(Cl)Cl
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?