• USA Home
  • 530743 - Lithium bis(dimethylethylsilyl)amide

EMAIL THIS PAGE TO A FRIEND
530743 Sigma-Aldrich

Lithium bis(dimethylethylsilyl)amide

99.99%

Purchase

Properties

assay   99.99%
form   liquid
bp   122 °C/0.2 mmHg (lit.)
density   0.892 g/mL at 25 °C (lit.)
SMILES string   [Li]N([Si](C)(C)CC)[Si](C)(C)CC
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1760 8 / PGII
WGK Germany 
3
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?