• USA Home
  • 530786 - Tributyltin isocyanate

EMAIL THIS PAGE TO A FRIEND
530786 Sigma-Aldrich

Tributyltin isocyanate

  • CAS Number 681-99-2

  • Linear Formula [CH3(CH2)3]3SnNCO

  • Molecular Weight 332.07

  •  MDL number MFCD08272321

  •  PubChem Substance ID 24877773

  •  NACRES NA.22

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotins
Quality Level   100
refractive index   n20/D 1.489 (lit.)
bp   297 °C (lit.)
density   1.203 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   CCCC[Sn](CCCC)(CCCC)N=C=O
InChI   1S/3C4H9.CNO.Sn/c3*1-3-4-2;2-1-3;/h3*1,3-4H2,2H3;;/q;;;-1;+1
InChI key   YGUNDMHXMXDGII-UHFFFAOYSA-N

Description

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2788 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
230.0 °F - closed cup
Flash Point(C) 
110 °C - closed cup
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?