• USA Home
  • 531839 - Dichloro[(R,R)-ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]zirconium(IV)

EMAIL THIS PAGE TO A FRIEND
531839 Sigma-Aldrich

Dichloro[(R,R)-ethylenebis(4,5,6,7-tetrahydro-1-indenyl)]zirconium(IV)

97%

  • CAS Number 109429-79-0

  • Empirical Formula (Hill Notation) C20H24Cl2Zr

  • Molecular Weight 426.53

  •  MDL number MFCD00672123

  •  PubChem Substance ID 24877844

  •  NACRES NA.22

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Other Chiral Catalysts
InChI Key   AKXWJOYQORUUHS-UHFFFAOYSA-L
assay   97%
optical activity   [α]20/D −89°, c = 1 in chloroform
mp   163-167 °C (lit.)

Description

Packaging

100 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?