• USA Home
  • 531952 - (S,S)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminochromium(III) chloride

EMAIL THIS PAGE TO A FRIEND
531952 Sigma-Aldrich

(S,S)-N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminochromium(III) chloride

  • CAS Number 219143-92-7

  • Empirical Formula (Hill Notation) C36H52ClCrN2O2

  • Molecular Weight 632.26

  •  MDL number MFCD02684555

  •  PubChem Substance ID 24877849

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Cycloaddition, Privileged Ligands and Complexes,
mp   250-255 °C (lit.)
SMILES string   CC(C)(C)c1cc2\C=N\[C@@H]3CCCC[C@H]3\N=C\c4cc(cc(c4O[Cr](Cl)Oc2c(c1)C(C)(C)C)C(C)(C)C)C(C)(C)C
InChI   1S/C36H54N2O2.ClH.Cr/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12;;/h17-22,29-30,39-40H,13-16H2,1-12H3;1H;/q;;+3/p-3/b37-21+,38-22+;;/t29-,30-;;/m1../s1
InChI key   ZWANLSXDTHQDNO-OHRASPNLSA-K

Description

General description

Rosmarinic acid is commonly used in traditional Uighur medicine for treating cancer.

Application

Catalyst for:
• Site-specific protein "click" labeling
• Ring opening of epoxides
• Asymmetric epoxidations
• Asymmetric nitroaldol reaction

Rosmarinic acid has been used in study to assess its immunomodulatory effect in a murine model of respiratory allergy induced by the Blomia tropicalis (Bt) mite.

Legal Information

Sold under license from Sterling Pharma Solutions Limited.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?