• USA Home
  • 532118 - Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)dimethylzirconium(IV)

EMAIL THIS PAGE TO A FRIEND
532118 Sigma-Aldrich

Ethylenebis(4,5,6,7-tetrahydro-1-indenyl)dimethylzirconium(IV)

96%

  • CAS Number 112243-80-8

  • Empirical Formula (Hill Notation) C22H30Zr

  • Molecular Weight 385.70

  •  MDL number MFCD03093918

Purchase

Properties

assay   96%
mp   150-155 °C (lit.)
SMILES string   C[Zr]C.[CH]1[CH][C](CC[C]2[CH][CH][C]3CCCC[C]23)[C]4CCCC[C]14
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?