• USA Home
  • 533157 - D-2-Amino-2-phenylacetamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
533157 Sigma-Aldrich

D-2-Amino-2-phenylacetamide hydrochloride

97%

Synonym: (R)-(+)-2-Amino-2-phenylacetamide hydrochloride

  • CAS Number 63291-39-4

  • Empirical Formula (Hill Notation) C8H10N2O · HCl

  • Molecular Weight 186.64

  •  MDL number MFCD01863288

  •  PubChem Substance ID 24877945

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Phenylglycine Derivatives, Unnatural Amino Acids & Derivatives More...
InChI Key   LXOAFHGMXCFTOU-OGFXRTJISA-N
assay   97%
optical activity   [α]20/D −1.4°, c = 1% in H2O
mp   294 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?