• USA Home
  • 533181 - 1,2-O-Cyclohexylidene-α-D-xylopentodialdo-1,4-furanose dimer

EMAIL THIS PAGE TO A FRIEND
533181 Sigma-Aldrich

1,2-O-Cyclohexylidene-α-D-xylopentodialdo-1,4-furanose dimer

97%

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
assay   97%
optical activity   [α]20/D +32.5°, c = 2 in chloroform
mp   187-191 °C (lit.)
SMILES string   O[C@@H](O[C@H]1[C@H](O[C@@H]2OC3(CCCCC3)O[C@H]12)C=O)[C@H]4O[C@@H]5OC6(CCCCC6)O[C@@H]5[C@H]4O
InChI   1S/C22H32O10/c23-11-12-14(17-20(26-12)32-22(30-17)9-5-2-6-10-22)27-18(25)15-13(24)16-19(28-15)31-21(29-16)7-3-1-4-8-21/h11-20,24-25H,1-10H2/t12-,13+,14+,15+,16-,17-,18?,19-,20-/m1/s1
InChI key   ZFTDWXNWWOYVDC-SPIWHNKXSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?