• USA Home
  • 533386 - Pentyltrichlorosilane

EMAIL THIS PAGE TO A FRIEND
533386 Sigma-Aldrich

Pentyltrichlorosilane

99%

Synonym: Amyltrichlorosilane

Purchase

Properties

InChI Key   KWDQAHIRKOXFAV-UHFFFAOYSA-N
assay   99%
form   liquid
refractive index   n20/D 1.4405 (lit.)
bp   173 °C(lit.)
density   1.142 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 2986 3(8) / PGII
WGK Germany 
1
RTECS 
VV4725000
Flash Point(F) 
87.8 °F
Flash Point(C) 
31 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?