• USA Home
  • 533483 - Trityl chloride, polymer-bound

EMAIL THIS PAGE TO A FRIEND
533483 Aldrich

Trityl chloride, polymer-bound

100-200 mesh, extent of labeling: 1.0-1.8 mmol/g loading, 1 % cross-linked

Synonym: Chlorotriphenylmethane, polymer-bound, Polystyrene crosslinked with divinylbenzene, α-chlorobenzhydrylated, Triphenylchloromethane, polymer-bound, Triphenylmethyl chloride

Purchase

Properties

Related Categories Chemical Synthesis, Other Supported Reagents, Solid Supported Synthesis, Specialty Synthesis, Supported Reagents More...
crosslinking   1 % cross-linked
extent of labeling   1.0-1.8 mmol/g loading
particle size   100-200 mesh
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?