• USA Home
  • 533742 - 5-(4-Methylphenyl)-1H-tetrazole

EMAIL THIS PAGE TO A FRIEND
533742 Sigma-Aldrich

5-(4-Methylphenyl)-1H-tetrazole

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Tetrazoles
InChI Key   BCCJIAZPYBJASR-UHFFFAOYSA-N
assay   97%
mp   250-253 °C (lit.)

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?