EMAIL THIS PAGE TO A FRIEND
535184 Sigma-Aldrich

Fmoc-Thr(tBu)-OH

97%

Synonym: N-Fmoc-O-tert-Butyl-L-threonine, Fmoc-O-tert-butyl-L-threonine

  • CAS Number 71989-35-0

  • Empirical Formula (Hill Notation) C23H27NO5

  • Molecular Weight 397.46

  •  Beilstein/REAXYS Number 4581133

  •  EC Number 276-261-1

  •  MDL number MFCD00077075

  •  PubChem Substance ID 24878078

Purchase

Properties

assay   97%
optical activity   [α]20/D +40°, c = 1 in chloroform
mp   124-128 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

47622 Fmoc-Thr(tBu)-OH, ≥98.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?