• USA Home
  • 535885 - 4-(2-Furyl)-1-buten-4-ol

EMAIL THIS PAGE TO A FRIEND
535885 Sigma-Aldrich

4-(2-Furyl)-1-buten-4-ol

97%

  • CAS Number 6398-51-2

  • Empirical Formula (Hill Notation) C8H10O2

  • Molecular Weight 138.16

  •  MDL number MFCD03093935

  •  PubChem Substance ID 24878113

Purchase

Properties

assay   97%
refractive index   n20/D 1.492 (lit.)
bp   204-205 °C (lit.)
density   1.06 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
2
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?