• USA Home
  • 535915 - 4-(4-Methoxyphenyl)-1-buten-4-ol

EMAIL THIS PAGE TO A FRIEND
535915 Sigma-Aldrich

4-(4-Methoxyphenyl)-1-buten-4-ol

97%

Purchase

Properties

assay   97%
refractive index   n20/D >1.5370 (lit.)
bp   279 °C(lit.)
density   1.047 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?