• USA Home
  • 536342 - 3-Propyl-5-hydroxy-5-D-arabinotetrahydroxybutyl-3-thiazolidine-2-thione

EMAIL THIS PAGE TO A FRIEND
536342 Aldrich

3-Propyl-5-hydroxy-5-D-arabinotetrahydroxybutyl-3-thiazolidine-2-thione

97%

  • CAS Number 63348-12-9

  • Empirical Formula (Hill Notation) C10H19NO5S2

  • Molecular Weight 297.39

  •  MDL number MFCD03093942

  •  PubChem Substance ID 24878153

Purchase

Properties

InChI Key   SJAKLSXXQWGQEG-UUBZBTQISA-N
assay   97%
optical activity   [α]20/D +160°, c = 2 in methanol
mp   177-181 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?