EMAIL THIS PAGE TO A FRIEND
53678 Sigma-Aldrich

γ-Humulene

technical, ~85% (GC)

Synonym: trans,trans-5-Methylene-1,8,8-trimethyl-1,6-cycloundecadiene, Isohumulene

  • CAS Number 26259-79-0

  • Empirical Formula (Hill Notation) C15H24

  • Molecular Weight 204.35

  •  Beilstein/REAXYS Number 1943966

  •  PubChem Substance ID 24878178

Purchase

Properties

grade   technical
assay   ~85% (GC)
refractive index   n20/D 1.515
density   0.9 g/mL at 20 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?