• USA Home
  • 538027 - Dimethylsiloxy-functionalized silica gel
EMAIL THIS PAGE TO A FRIEND
538027 Sigma-Aldrich
Dimethylsiloxy-functionalized silica gel
200-400 mesh, extent of labeling: 1.5 mmol/g loading
Purchase
Properties
extent of labeling   1.5 mmol/g loading
particle size   200-400 mesh
pore size   60 Å pore size
surface area   500 m2/g
Safety & Documentation
Safety Information
Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?