• USA Home
  • 538043 - 3-(Dimethylamino)propyl-functionalized silica gel

EMAIL THIS PAGE TO A FRIEND
538043 Sigma-Aldrich

3-(Dimethylamino)propyl-functionalized silica gel

200-400 mesh, extent of labeling: 1.4 mmol/g loading

Purchase

Properties

extent of labeling   1.4 mmol/g loading
particle size   200-400 mesh
pore size   60 Å pore size
surface area   500 m2/g
SMILES string   O=[SiH2].CCCN(C)C
InChI   1S/C5H13N.H2OSi/c1-4-5-6(2)3;1-2/h4-5H2,1-3H3;2H2
InChI key   UMCMWYYBIGZTGT-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Functionalized Silica Gel Scavengers

Sigma-Aldrich Corporation and SiliCycle Inc. have collaborated to offer scientists ultra pure functionalized silica gels for medicinal and parallel chemistry. These silica gels offer several advantag...
Chemfiles Volume 3 Article 5
Keywords: Microwave synthesis, Organic synthesis, Parallel synthesis, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?