• USA Home
  • 538272 - 2-(2-Ethylhexyloxy)ethanol

EMAIL THIS PAGE TO A FRIEND
538272 Sigma-Aldrich

2-(2-Ethylhexyloxy)ethanol

82%

Synonym: EEH Solvent, Ethylene glycol 2-ethylhexyl ether

  • CAS Number 1559-35-9

  • Linear Formula CH3(CH2)3CH(C2H5)CH2OCH2CH2OH

  • Molecular Weight 174.28

  •  MDL number MFCD00020602

  •  PubChem Substance ID 24878316

Purchase

Properties

InChI Key   OHJYHAOODFPJOD-UHFFFAOYSA-N
assay   82%
refractive index   n20/D 1.435(lit.)
bp   229 °C(lit.)
density   0.892 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?