• USA Home
  • 538949 - Bis(2-methyl-8-hydroxyquinolinato)zinc

EMAIL THIS PAGE TO A FRIEND
538949 Sigma-Aldrich

Bis(2-methyl-8-hydroxyquinolinato)zinc

98%

  • CAS Number 14128-73-5

  • Empirical Formula (Hill Notation) C20H16N2O2Zn

  • Molecular Weight 381.74

  •  MDL number MFCD03093700

  •  PubChem Substance ID 24878368

Purchase

Properties

assay   98%
mp   285-290 °C (lit.)
fluorescence   λex 266 nm; λem 554 nm in acetonitrile

Description

General description

Light-emitting metal complex

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?