• USA Home
  • 538981 - (R)-(−)-1,3-Nonanediol

EMAIL THIS PAGE TO A FRIEND
538981 Sigma-Aldrich

(R)-(−)-1,3-Nonanediol

97%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   CGNJFUJNEYIYRZ-SECBINFHSA-N
assay   97%
refractive index   n20/D >1.4510 (lit.)
bp   271-272 °C(lit.)
density   >0.929 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?