• USA Home
  • 539007 - (R)-(+)-1-(4-Methoxybenzyl)-1,2,3,4,5,6,7,8-octahydroisoquinoline

EMAIL THIS PAGE TO A FRIEND
539007 Sigma-Aldrich

(R)-(+)-1-(4-Methoxybenzyl)-1,2,3,4,5,6,7,8-octahydroisoquinoline

97%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Isoquinolines More...
assay   97%
optical activity   [α]20/D +152°, c = 1 in chloroform
refractive index   n20/D 1.565 (lit.)
density   1.108 g/mL at 25 °C (lit.)
SMILES string   COc1ccc(C[C@H]2NCCC3=C2CCCC3)cc1
InChI   1S/C17H23NO/c1-19-15-8-6-13(7-9-15)12-17-16-5-3-2-4-14(16)10-11-18-17/h6-9,17-18H,2-5,10-12H2,1H3/t17-/m1/s1
InChI key   NPEVCJZMQGZNET-QGZVFWFLSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?