• USA Home
  • 539465 - N,N-Dimethylformamide

EMAIL THIS PAGE TO A FRIEND
539465 Sigma-Aldrich

N,N-Dimethylformamide

AldraSORB, 99.8%

Synonym: DMF, NSC 5356

  • CAS Number 68-12-2

  • Linear Formula HCON(CH3)2

  • Molecular Weight 73.09

  •  Beilstein/REAXYS Number 605365

  •  EC Number 200-679-5

  •  MDL number MFCD00003284

  •  PubChem Substance ID 24878402

Purchase

Properties

Related Categories Anhydrous Solvents, Dimethylformamide (DMF), Solvent by Type, Solvent by Application, Solvents More...
vapor density   2.5 (vs air)
vapor pressure   2.7 mmHg ( 20 °C)
InChI Key   ZMXDDKWLCZADIW-UHFFFAOYSA-N
assay   99.8%
autoignition temp.   833 °F
expl. lim.   15.2 %
impurities   <0.005% water
evapn. residue   <0.0005%
refractive index   n20/D 1.430 (lit.)
bp   153 °C (lit.)
mp   −61 °C (lit.)
density   0.944 g/mL (lit.)

Description

Application

Solvent for many hydrophobic organic compounds.

Legal Information

AldraSORB is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2265 3 / PGIII
WGK Germany 
2
RTECS 
LQ2100000
Flash Point(F) 
136.4 °F
Flash Point(C) 
58 °C

Dispensette™ S
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Products

comparable product

Product #

Description

Add to Cart

227056 N,N-Dimethylformamide, anhydrous, 99.8%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?