• USA Home
  • 539562 - 2-Fluoro-6-(trifluoromethyl)benzyl alcohol

EMAIL THIS PAGE TO A FRIEND
539562 Sigma-Aldrich

2-Fluoro-6-(trifluoromethyl)benzyl alcohol

99%

Purchase

Properties

assay   99%
refractive index   n20/D 1.456 (lit.)
bp   180 °C (lit.)
density   1.397 g/mL at 25 °C (lit.)
SMILES string   OCc1c(F)cccc1C(F)(F)F
InChI   1S/C8H6F4O/c9-7-3-1-2-6(5(7)4-13)8(10,11)12/h1-3,13H,4H2
InChI key   TZUCYZIRXZYZOP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?