• USA Home
  • 539619 - 2-Fluoro-6-(trifluoromethyl)benzamide

EMAIL THIS PAGE TO A FRIEND
539619 Sigma-Aldrich

2-Fluoro-6-(trifluoromethyl)benzamide

98%

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
assay   98%
mp   144-147 °C (lit.)
SMILES string   NC(=O)c1c(F)cccc1C(F)(F)F
InChI   1S/C8H5F4NO/c9-5-3-1-2-4(8(10,11)12)6(5)7(13)14/h1-3H,(H2,13,14)
InChI key   BYQGJCJJFXIYHC-UHFFFAOYSA-N

Description

General description

2-Fluoro-6-(trifluoromethyl)benzamide, also known as 6-fluoro-2-(trifluoromethyl)benzamide, is a fluorinated aromatic amide.

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?