• USA Home
  • 54514 - (2-Hydroxyethyl)triphenylphosphonium chloride

EMAIL THIS PAGE TO A FRIEND
54514 Sigma-Aldrich

(2-Hydroxyethyl)triphenylphosphonium chloride

≥98.0%

  • CAS Number 23250-03-5

  • Linear Formula HOCH2CH2P(C6H5)3Cl

  • Molecular Weight 342.80

  •  Beilstein/REAXYS Number 3922778

  •  EC Number 245-521-6

  •  MDL number MFCD00011754

  •  PubChem Substance ID 24878811

  •  NACRES NA.22

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Olefination, Synthetic Reagents, Wittig Reagents More...
assay   ≥98.0%
reaction suitability   reaction type: C-C Bond Formation
mp   234-235 °C (dec.) (lit.)
  238-240 °C (dec.)
storage temp.   room temp
SMILES string   [Cl-].OCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
InChI   1S/C20H20OP.ClH/c21-16-17-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20;/h1-15,21H,16-17H2;1H/q+1;/p-1
InChI key   NOEABYSOSUWXKG-UHFFFAOYSA-M

Description

Application

Reagent for:
• Wittig reaction

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?