• USA Home
  • 545317 - N,N-Dibenzyl-D-serine methyl ester

EMAIL THIS PAGE TO A FRIEND
545317 Sigma-Aldrich

N,N-Dibenzyl-D-serine methyl ester

97%

  • CAS Number 202478-34-0

  • Linear Formula HOCH2CH(CO2CH3)N(CH2C6H5)2

  • Molecular Weight 299.36

  •  MDL number MFCD03427072

Purchase

Properties

assay   97%
optical activity   [α]20/D +85°, c = 1% in methanol
refractive index   n20/D 1.5590 (lit.)
density   1.201 g/mL at 25 °C (lit.)
SMILES string   COC(=O)[C@@H](CO)N(Cc1ccccc1)Cc2ccccc2
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?