• USA Home
  • 545597 - N-(-3-Acetylthiopropyl)phthalimide

EMAIL THIS PAGE TO A FRIEND
545597 Sigma-Aldrich

N-(-3-Acetylthiopropyl)phthalimide

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Protected Thiols/Mercaptans, Sulfur Compounds More...
assay   97%
mp   90-94 °C (lit.)
SMILES string   CC(=O)SCCCN1C(=O)c2ccccc2C1=O
InChI   1S/C13H13NO3S/c1-9(15)18-8-4-7-14-12(16)10-5-2-3-6-11(10)13(14)17/h2-3,5-6H,4,7-8H2,1H3
InChI key   QDCSBRSBDZXURK-UHFFFAOYSA-N

Description

General description

N-(-3-Acetylthiopropyl)phthalimide is an N-substituted phthalimide derivative. It is also known as S-[3-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)propyl] ethanethioate.

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?