• USA Home
  • 546364 - 1-(5-Bromo-2-hydroxyphenyl)-1,3-butanedione

EMAIL THIS PAGE TO A FRIEND
546364 Sigma-Aldrich

1-(5-Bromo-2-hydroxyphenyl)-1,3-butanedione

  • CAS Number 207387-68-6

  • Linear Formula BrC6H3(OH)COCH2COCH3

  • Molecular Weight 257.08

  •  MDL number MFCD04039883

  •  PubChem Substance ID 24878891

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Chemical Synthesis, Ketones,
mp   113-118 °C (lit.)
SMILES string   CC(=O)CC(=O)c1cc(Br)ccc1O
InChI   1S/C10H9BrO3/c1-6(12)4-10(14)8-5-7(11)2-3-9(8)13/h2-3,5,13H,4H2,1H3
InChI key   GCPQDUDCTCGTCZ-UHFFFAOYSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?