• USA Home
  • 546720 - (R)-(+)-2-Methylsuccinic acid 4-methyl ester

EMAIL THIS PAGE TO A FRIEND
546720 Aldrich

(R)-(+)-2-Methylsuccinic acid 4-methyl ester

95%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
InChI Key   UVQYBUYGFBXQGO-SCSAIBSYSA-N
assay   95%
optical activity   [α]20/D +11°, c = 1 in chloroform
refractive index   n20/D 1.4320(lit.)
bp   241-242 °C/760 mmHg(lit.)
density   1.150 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
100.4 °F
Flash Point(C) 
38 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?