• USA Home
  • 547530 - StratoSpheres PL-NCO (Isocyanate) resin

EMAIL THIS PAGE TO A FRIEND
547530 Sigma-Aldrich

StratoSpheres PL-NCO (Isocyanate) resin

150-300 μm, extent of labeling: ≥1.5 mmol/g loading, 1 % cross-linked

Synonym: Isocyanate, polymer-bound, Methyl isocyanate bound to polystyrene

  • CAS Number 59990-69-1

  • Empirical Formula (Hill Notation) C26H25NO

  • Molecular Weight 367.48

  •  MDL number MFCD01867272

  •  PubChem Substance ID 24878971

  •  NACRES NA.22

Purchase

Properties

Related Categories Chemical Synthesis, Solid Supported Synthesis, Specialty Synthesis, StratoSpheres
crosslinking   1 % cross-linked
extent of labeling   ≥1.5 mmol/g loading
particle size   150-300 μm
storage temp.   room temp

Description

Packaging

5 g in glass bottle

Legal Information

StratoSpheres is a trademark of Agilent Technologies, Inc.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?