EMAIL THIS PAGE TO A FRIEND
548537 Sigma-Aldrich

Methanol-d4

99.8 atom % D

Synonym: Methyl-d3 alcohol d, Tetradeuteromethanol

Purchase

Properties

isotopic purity   99.8 atom % D
impurities   ≤0.025% water
refractive index   n20/D 1.326 (lit.)
bp   65.4 °C (lit.)
density   0.888 g/mL at 25 °C (lit.)
SMILES string   [2H]OC([2H])([2H])[2H]
InChI   1S/CH4O/c1-2/h2H,1H3/i1D3,2D
InChI key   OKKJLVBELUTLKV-MZCSYVLQSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
WGK 2
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?