EMAIL THIS PAGE TO A FRIEND
548561 Aldrich

Sodium pyrovanadate

98%

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

RIDADR 
UN 3285 6.1 / PGIII
WGK Germany 
3
RTECS 
WD7900000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?