• USA Home
  • 550108 - Methyl (6-methyl-2H-1,4-benzoxazin-3(4H)-one-2-yl)acetate

EMAIL THIS PAGE TO A FRIEND
550108 Sigma-Aldrich

Methyl (6-methyl-2H-1,4-benzoxazin-3(4H)-one-2-yl)acetate

97%

Purchase

Properties

Related Categories Benzoxazines, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
assay   97%
mp   163-167 °C (lit.)
SMILES string   COC(=O)CC1Oc2ccc(C)cc2NC1=O
InChI   1S/C12H13NO4/c1-7-3-4-9-8(5-7)13-12(15)10(17-9)6-11(14)16-2/h3-5,10H,6H2,1-2H3,(H,13,15)
InChI key   GSPIARUBRADJEA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?