• USA Home
  • 550922 - 1-(2-Chlorophenyl)-1-propanol

EMAIL THIS PAGE TO A FRIEND
550922 Sigma-Aldrich

1-(2-Chlorophenyl)-1-propanol

95%

Purchase

Properties

assay   95%
refractive index   n20/D >1.5390 (lit.)
bp   248-249 °C (lit.)
density   1.145 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?