• USA Home
  • 55105 - 8-Hydroxy-2-quinolinesulfonic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
55105 Aldrich

8-Hydroxy-2-quinolinesulfonic acid monohydrate

≥97.0%

  • CAS Number 20946-17-2

  • Empirical Formula (Hill Notation) C9H7NO4S · H2O

  • Molecular Weight 243.24

  •  Beilstein/REAXYS Number 401756

  •  MDL number MFCD00216718

  •  PubChem Substance ID 24879191

Purchase

Properties

assay   ≥97.0%
mp   >300 °C (lit.)
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?