• USA Home
  • 551619 - Manganese(II) iodide

EMAIL THIS PAGE TO A FRIEND
551619 Sigma-Aldrich

Manganese(II) iodide

powder, anhydrous, 99.9%

Purchase

Properties

Related Categories Crystal Grade Inorganics, Essential Chemicals, Manganese, Manganese Salts, Materials Science,
grade   anhydrous
InChI Key   QWYFOIJABGVEFP-UHFFFAOYSA-L
assay   99.9%
form   powder
mp   80 °C (dec.) (lit.)
density   5.01 g/mL at 25 °C(lit.)
Featured Industry   Battery Manufacturing
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Lithium-Ion Battery Performance: Dependence on Material Synthesis and Post‑Treatment Methods

Dr. Qifeng Zhang, Mr. Evan Uchaker, and Dr. Guozhong Cao* Department of Materials Science and Engineering University of Washington Seattle, WA 98195-2120 *Email: gzcao@u.washington.edu
Keywords: Absorption, Deposition, Diffraction, Diffusion, Electrochemical reactions, Materials Science, Nanomaterials, Nanotubes, Nucleic acid annealing, Oxidations, Precipitation, Redox Reactions, Reductions, X-Ray diffraction

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

related product

Product #

Description

Add to Cart

43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?