• USA Home
  • 552380 - (4-Amino-5-chloro-2-methoxy)-N-[3-methoxy(4-piperidyl)]benzamide

EMAIL THIS PAGE TO A FRIEND
552380 Sigma-Aldrich

(4-Amino-5-chloro-2-methoxy)-N-[3-methoxy(4-piperidyl)]benzamide

97%

Purchase

Properties

assay   97%
mp   190-193 °C (lit.)
SMILES string   COC1CNCCC1NC(=O)c2cc(Cl)c(N)cc2OC
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?