• USA Home
  • 552593 - 2-(4-Toluenesulfonyl hydrazino)ethyl-functionalized silica gel

EMAIL THIS PAGE TO A FRIEND
552593 Sigma-Aldrich

2-(4-Toluenesulfonyl hydrazino)ethyl-functionalized silica gel

200-400 mesh, extent of labeling: 1 mmol/g loading

Synonym: Sulfohydrazine-2ar functionalized silica gel

Purchase

Properties

extent of labeling   1 mmol/g loading
particle size   200-400 mesh
pore size   60 Å pore size
surface area   500 m2/g
mp   >330 °C (lit.)

Description

Application

Scavenges aldehydes and ketones.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Functionalized Silica Gel Scavengers

Sigma-Aldrich Corporation and SiliCycle Inc. have collaborated to offer scientists ultra pure functionalized silica gels for medicinal and parallel chemistry. These silica gels offer several advantag...
Chemfiles Volume 3 Article 5
Keywords: Microwave synthesis, Organic synthesis, Parallel synthesis, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?