EMAIL THIS PAGE TO A FRIEND
552992 Sigma-Aldrich

Tellurium dioxide

optical grade

Purchase

Properties

Related Categories Materials Science, Metal and Ceramic Science, Oxides, Tellurium
grade   optical grade
InChI Key   LAJZODKXOMJMPK-UHFFFAOYSA-N
mp   733 °C (lit.)
density   5.67 g/mL at 25 °C(lit.)

Description

Packaging

25 g in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WY2675000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?