EMAIL THIS PAGE TO A FRIEND
553018 Sigma-Aldrich

Zinc selenide

random pieces, optical grade

Purchase

Properties

grade   deposition grade
  optical grade
InChI Key   SBIBMFFZSBJNJF-UHFFFAOYSA-N
form   random pieces
density   5.42 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3283 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?