• USA Home
  • 554014 - Tris(cyclopentadienyl)holmium(III)

EMAIL THIS PAGE TO A FRIEND
554014 Sigma-Aldrich

Tris(cyclopentadienyl)holmium(III)

99.9%

Purchase

Properties

assay   99.9%
form   solid
mp   252 °C (dec.) (lit.)
SMILES string   [Ho].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.[CH]3[CH][CH][CH][CH]3
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3395 4.3 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?