EMAIL THIS PAGE TO A FRIEND
554294 Sigma-Aldrich

N-Z-D-Leucinol

97%

Synonym: N-Benzyloxycarbonyl-D-leucinol

  • CAS Number 166735-51-9

  • Linear Formula (CH3)2CHCH2CH(CH2OH)NHCO2CH2C6H5

  • Molecular Weight 251.32

  •  MDL number MFCD03427144

  •  PubChem Substance ID 24879389

  •  NACRES NA.22

Purchase

Properties

Related Categories Amino Acids & Derivatives, Amino Alcohols, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry More...
Quality Level   100
assay   97%
optical activity   [α]20/D +27°, c = 1% in chloroform
application(s)   peptide synthesis: suitable
mp   30-34 °C (lit.)
SMILES string   CC(C)C[C@H](CO)NC(=O)OCc1ccccc1
InChI   1S/C14H21NO3/c1-11(2)8-13(9-16)15-14(17)18-10-12-6-4-3-5-7-12/h3-7,11,13,16H,8-10H2,1-2H3,(H,15,17)/t13-/m1/s1
InChI key   YGARWTHXCINBQM-CYBMUJFWSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?