• USA Home
  • 55485 - Hydroxylamine phosphate

EMAIL THIS PAGE TO A FRIEND
55485 Sigma-Aldrich

Hydroxylamine phosphate

≥95.0% (RT)

Synonym: Hydroxylammonium phosphate

Purchase

Properties

assay   ≥95.0% (RT)
impurities   ≤2% water
SMILES string   NO.NO.NO.OP(O)(O)=O
InChI   1S/3H3NO.H3O4P/c3*1-2;1-5(2,3)4/h3*2H,1H2;(H3,1,2,3,4)
InChI key   XBUFCZMOAHHGMX-UHFFFAOYSA-N

Description

Application

Reactant for:
• Aqueous preparation of rhenium diphosphate
• Gas and solid state reactions with acetone
• Hydroxylamine formation via photolysis

Safety & Documentation

Safety Information

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?