EMAIL THIS PAGE TO A FRIEND
554952 Sigma-Aldrich

Isoamyl sulfide

≥98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfides/Disulfides, Sulfur Compounds More...
assay   ≥98%
refractive index   n20/D 1.453 (lit.)
bp   49 °C/0.1 mmHg (lit.)
density   0.834 g/mL at 25 °C (lit.)
storage temp.   room temp
SMILES string   CC(C)CCSCCC(C)C
InChI   1S/C10H22S/c1-9(2)5-7-11-8-6-10(3)4/h9-10H,5-8H2,1-4H3
InChI key   JWEWNTJADCWFRP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
183.2 °F
Flash Point(C) 
84 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?