• USA Home
  • 554979 - Poly[9,9-di(3′,7′-dimethyloctyl)fluoren-2,7-yleneethynylene]

EMAIL THIS PAGE TO A FRIEND
554979 Sigma-Aldrich

Poly[9,9-di(3′,7′-dimethyloctyl)fluoren-2,7-yleneethynylene]

Purchase

Properties

Related Categories Light-Emitting Polymers (LEPs), Materials Science, OLED and PLED Materials, Organic and Printed Electronics, Poly(fluorenylene ethynylene) (PFE) Polymers More...
mp   110 °C (dec.) (lit.)
solubility   chlorinated solvents: soluble (including chloroform)
λmax   381 nm
fluorescence   λex 385 nm; λem 419 nm in chloroform

Description

Packaging

500 mg in glass insert

Application

Conducting polymer. Exhibits blue photo- and electroluminescence.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?