• USA Home
  • 555517 - 4-(Trifluoromethylthio)phenylacetonitrile

EMAIL THIS PAGE TO A FRIEND
555517 Sigma-Aldrich

4-(Trifluoromethylthio)phenylacetonitrile

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.490 (lit.)
density   1.331 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?