• USA Home
  • 557064 - Tetraphenylantimony(V) acetate

EMAIL THIS PAGE TO A FRIEND
557064 Sigma-Aldrich

Tetraphenylantimony(V) acetate

98%

Purchase

Properties

Related Categories Antimony Catalysts, Catalysis and Inorganic Chemistry, Chemical Synthesis More...
InChI Key   MOQVJYRFLPHSMW-UHFFFAOYSA-M
assay   98%
mp   120-133 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?