EMAIL THIS PAGE TO A FRIEND
557951 Sigma-Aldrich

(R)-(−)-2-Nonanol

97%

Purchase

Properties

assay   97%
optical activity   [α]20/D -8.50°, c = 1% in chloroform
optical purity   ee: 98% (GLC)
mp   50-53 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NA 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
179.6 °F
Flash Point(C) 
82 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?